Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-L-phenylalanine, 98%

$159.72 - $399.30

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-L-phenylalanine
CAS 31105-91-6
Molecular Formula C9H9F2NO2
Formula Weight 201.18
MDL Number MFCD01631987
Synonym H-Phe(3,5-DiF)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2783303

  SDS
alfa aesar™
H2783303
1g Each for $399.30

AAH27833MD

  SDS
alfa aesar™
H27833MD
250mg Each for $159.72
Description & Specifications

Specifications

Formula Weight 201.18
Chemical Name or Material 3,5-Difluoro-L-phenylalanine
Assay 98%
Molecular Formula C9H9F2NO2
MDL Number MFCD01631987
CAS 31105-91-6
Synonym H-Phe(3,5-DiF)-OH
Quantity 1g