Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-D-phenylalanine, 98%

$210.39 - $210.39

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-D-phenylalanine
CAS 266360-63-8
Molecular Formula C9H9F2NO2
Formula Weight 201.18g/mol
MDL Number MFCD01631988
Synonym H-D-Phe(3,5-DiF)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2700703

 
alfa aesar™
H27007-03
1g Each for $210.39
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 3,5-Difluoro-D-phenylalanine
CAS 266360-63-8
Assay 98%
Molecular Formula C9H9F2NO2
Formula Weight 201.18g/mol
MDL Number MFCD01631988
Synonym H-D-Phe(3,5-DiF)-OH
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.