Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-4-methoxyphenol, 97%

$230.89 - $230.89

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-4-methoxyphenol
CAS 443-42-5
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD04115918
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2645103

 
alfa aesar™
H2645103
1g Each for $230.89
Description & Specifications

Specifications

Formula Weight 160.12
Chemical Name or Material 3,5-Difluoro-4-methoxyphenol
Assay 97%
Molecular Formula C7H6F2O2
MDL Number MFCD04115918
CAS 443-42-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 67° to 70°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.