Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester
CAS 890839-37-9
Formula Weight 270.08
Synonym 3,5-Difluoro-4-methoxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6257703

 
alfa aesar™
H6257703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62577MD

 
alfa aesar™
H62577MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 270.08
Chemical Name or Material 3,5-Difluoro-4-methoxybenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
CAS 890839-37-9
Synonym 3,5-Difluoro-4-methoxyphenylboronic acid pinacol ester
Quantity 1g