Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-4-methoxybenzaldehyde, 98%

$100.97 - $306.31

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-4-methoxybenzaldehyde
CAS 654-11-5
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD04115911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26219MC

  SDS
alfa aesar™
H26219MC
100mg Each for $100.97

AAH26219ME

  SDS
alfa aesar™
H26219ME
500mg Each for $306.31
Description & Specifications

Specifications

Formula Weight 172.13
Chemical Name or Material 3,5-Difluoro-4-methoxybenzaldehyde
Assay 98%
Molecular Formula C8H6F2O2
MDL Number MFCD04115911
CAS 654-11-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 38° to 40°C
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.