Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',5'-Difluoro-4'-hydroxypropiophenone, 98%

$450.53 - $450.53

Chemical Identifiers

Chemical Name or Material 3',5'-Difluoro-4'-hydroxypropiophenone
CAS 178374-78-2
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD00061287
Beilstein 7372089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2022906

  SDS
alfa aesar™
B2022906
5g Each for $450.53
Description & Specifications

Specifications

Formula Weight 186.16
Melting Point 128° to 132°C
Assay 98%
Health Hazard 1 Warning
Beilstein 7372089
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
CAS 178374-78-2
Molecular Formula C9H8F2O2
Chemical Name or Material 3',5'-Difluoro-4'-hydroxypropiophenone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00061287
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.