Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Difluoro-2-methylbenzaldehyde, 97%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 3,5-Difluoro-2-methylbenzaldehyde
Molecular Formula C8H6F2O
Formula Weight 156.13
MDL Number MFCD09832335
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3185103

 
alfa aesar™
H3185103
1g Each for $213.96
Description & Specifications

Specifications

Formula Weight 156.13
Chemical Name or Material 3,5-Difluoro-2-methylbenzaldehyde
Assay 97%
Molecular Formula C8H6F2O
MDL Number MFCD09832335
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 32° to 35°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.