Learn More
(3,5-Difluoro-2-methoxyphenyl)boronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC365820050
Specifications
(3, 5-Difluoro-2-methoxyphenyl)boronic acid | |
Glass bottle | |
200°C | |
97% | |
96.0 | |
96% min. (HPLC) | |
MFCD03095353 | |
JXQHRWOUVJRCSR-UHFFFAOYSA-N | |
(3,5-difluoro-2-methoxyphenyl)boronic acid | |
2734607 | |
Powder and Chunks |
Authentic | |
Beige | |
5g | |
737000-76-9 | |
100.0 | |
C7H7BF2O3 | |
2-methoxy-3,5-difluoro-benzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxyphenyl boronicacid, acmc-209orz, b-3,5-difluoro-2-methoxyphenyl-boronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl | |
B(C1=CC(=CC(=C1OC)F)F)(O)O | |
187.81 | |
187.81 | |
97% |
Chemical Identifiers
737000-76-9 | |
187.81 | |
JXQHRWOUVJRCSR-UHFFFAOYSA-N | |
2734607 | |
B(C1=CC(=CC(=C1OC)F)F)(O)O |
C7H7BF2O3 | |
MFCD03095353 | |
2-methoxy-3,5-difluoro-benzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxyphenyl boronicacid, acmc-209orz, b-3,5-difluoro-2-methoxyphenyl-boronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl | |
(3,5-difluoro-2-methoxyphenyl)boronic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning