Please login to your online account to display your discounted pricing

(3,5-Difluoro-2-methoxyphenyl)boronic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365820010

 View more versions of this product

Catalog No. AC365820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder and Chunks
Formula Weight 187.81
Quantity 1g
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (3, 5-Difluoro-2-methoxyphenyl)boronic acid
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H7BF2O3
MDL Number MFCD03095353
CAS 737000-76-9
Synonym 2-Methoxy-3, 5-difluorophenylboronic acid
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (3, 5-Difluoro-2-methoxyphenyl)boronic acid
Name Note 97%
CAS 737000-76-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7BF2O3
Formula Weight 187.81
MDL Number MFCD03095353
Synonym 2-Methoxy-3, 5-difluorophenylboronic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents