Learn More
3,5-Difluoro-2-methoxybenzyl bromide, 97%, Thermo Scientific™
$235.16 - $702.27
Chemical Identifiers
CAS | 886500-63-6 |
---|---|
Molecular Formula | C8H7BrF2O |
Molecular Weight (g/mol) | 237.04 |
MDL Number | MFCD06660240 |
InChI Key | XLYRQPGSEJMVIM-UHFFFAOYSA-N |
Synonym | 1-bromomethyl-3,5-difluoro-2-methoxybenzene, 3,5-difluoro-2-methoxybenzyl bromide, 3,5-difluoro-2-methoxybenzylbromide, benzene,1-bromomethyl-3,5-difluoro-2-methoxy, 1-bromomethyl-3,5-bis fluoranyl-2-methoxy-benzene |
PubChem CID | 17750690 |
IUPAC Name | 1-(bromomethyl)-3,5-difluoro-2-methoxybenzene |
SMILES | COC1=C(F)C=C(F)C=C1CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2658203
|
Thermo Scientific Chemicals
H2658203 |
1 g |
Each for $235.16
|
|
|||||
AAH2658206
|
Thermo Scientific Chemicals
H2658206 |
5 g |
Each for $702.27
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
886500-63-6 | |
237.04 | |
XLYRQPGSEJMVIM-UHFFFAOYSA-N | |
17750690 | |
COC1=C(F)C=C(F)C=C1CBr |
C8H7BrF2O | |
MFCD06660240 | |
1-bromomethyl-3,5-difluoro-2-methoxybenzene, 3,5-difluoro-2-methoxybenzyl bromide, 3,5-difluoro-2-methoxybenzylbromide, benzene,1-bromomethyl-3,5-difluoro-2-methoxy, 1-bromomethyl-3,5-bis fluoranyl-2-methoxy-benzene | |
1-(bromomethyl)-3,5-difluoro-2-methoxybenzene |
Specifications
886500-63-6 | |
MFCD06660240 | |
UN3265 | |
XLYRQPGSEJMVIM-UHFFFAOYSA-N | |
1-(bromomethyl)-3,5-difluoro-2-methoxybenzene | |
17750690 | |
97% |
C8H7BrF2O | |
1 g | |
1-bromomethyl-3,5-difluoro-2-methoxybenzene, 3,5-difluoro-2-methoxybenzyl bromide, 3,5-difluoro-2-methoxybenzylbromide, benzene,1-bromomethyl-3,5-difluoro-2-methoxy, 1-bromomethyl-3,5-bis fluoranyl-2-methoxy-benzene | |
COC1=C(F)C=C(F)C=C1CBr | |
237.04 | |
237.04 | |
3,5-Difluoro-2-methoxybenzyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures