Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, tech. 75%

$172.41 - $566.46

Chemical Identifiers

Chemical Name or Material 3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine
CAS 34562-31-7
Purity Grade Technical Grade
Molecular Formula C18H25N
Formula Weight 255.4
MDL Number MFCD00071789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3253809

  SDS
alfa aesar™
H3253809
10g Each for $172.41

AAH3253818

  SDS
alfa aesar™
H3253818
50g Each for $566.46
Description & Specifications

Specifications

Formula Weight 255.4
Chemical Name or Material 3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine
TSCA TSCA
Purity Grade Technical Grade
Assay 75%
Molecular Formula C18H25N
MDL Number MFCD00071789
CAS 34562-31-7
Boiling Point 125°C (0.5mmHg)
Flash Point >100°C (212°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.