Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorothiophenol, 97%

$85.14 - $308.89

Chemical Identifiers

Chemical Name or Material 3,5-Dichlorothiophenol
CAS 17231-94-6
Molecular Formula C6H4Cl2S
Formula Weight 179.07
MDL Number MFCD00041423
Beilstein 2041184
Synonym 3,5-Dichlorobenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1069703

  SDS
alfa aesar™
L1069703
1g Each for $85.14

AAL1069706

  SDS
alfa aesar™
L1069706
5g Each for $308.89
Description & Specifications

Specifications

Formula Weight 179.07
CAS 17231-94-6
Chemical Name or Material 3,5-Dichlorothiophenol
MDL Number MFCD00041423
Molecular Formula C6H4Cl2S
Synonym 3,5-Dichlorobenzenethiol
Odor Unpleasant
Assay 97%
Health Hazard 1 Warning
Beilstein 2041184
Quantity 1g
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 60° to 63°C
Boiling Point 270° to 272°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.