missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorothiophenol, 97%

Manufacturer:  Alfa Aesar™ L1069703

 View more versions of this product

Catalog No. AAL1069703



Specifications

Specifications

3,5-Dichlorothiophenol
270°C to 272°C
1g
C6H4Cl2S
2041184
WRXIPCQPHZMXOO-UHFFFAOYSA-N
3,5-dichlorobenzenethiol
2736096
97%
Unpleasant
60°C to 63°C
17231-94-6
MFCD00041423
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
C1=C(C=C(C=C1Cl)Cl)S
179.058
179.07

Chemical Identifiers

17231-94-6
179.058
WRXIPCQPHZMXOO-UHFFFAOYSA-N
2736096
C1=C(C=C(C=C1Cl)Cl)S
C6H4Cl2S
MFCD00041423
3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
3,5-dichlorobenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents