missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorosalicylaldehyde, +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC407000250

 View more versions of this product

Catalog No. AC407000250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3, 5-Dichlorosalicylaldehyde
91° to 94°C
90-60-8
98.0
Glass bottle
≥98.0%
C7H4Cl2O2
MFCD00003320
Pale yellow
25g
Authentic
100.0
Crystalline Powder
98+%
191.01

Chemical Identifiers

90-60-8
100.0
C7H4Cl2O2
MFCD00003320
≥98.0%
98.0
98+%
191.01
3, 5-Dichlorosalicylaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning