Please login to your online account to display your discounted pricing

3,5-Dichloropyridine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dichloropyridine
Assay Percent Range 97%
Molecular Formula C5H3Cl2N
Linear Formula (C5H3N)Cl2
Formula Weight 147.99
MDL Number MFCD00006376
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC113741000

  SDS
thermo fisher scientific
AC113741000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.99
Quantity 100g
EINECS Number 219-537-9
Chemical Name or Material 3, 5-Dichloropyridine
CAS 2457-47-8
Linear Formula (C5H3N)Cl2
Molecular Formula C5H3Cl2N
Assay Percent Range 97%
MDL Number MFCD00006376