Please login to your online account to display your discounted pricing

3,5-Dichloropyridine, 97%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC113741000

 View more versions of this product

Catalog No. AC113741000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 147.99
Quantity 100g
EINECS Number 219-537-9
Chemical Name or Material 3, 5-Dichloropyridine
CAS 2457-47-8
Linear Formula (C5H3N)Cl2
Molecular Formula C5H3Cl2N
Assay Percent Range 97%
MDL Number MFCD00006376

Specifications

Chemical Name or Material 3, 5-Dichloropyridine
Assay Percent Range 97%
Molecular Formula C5H3Cl2N
Linear Formula (C5H3N)Cl2
Formula Weight 147.99
MDL Number MFCD00006376