Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloropyridine-2-carboxamide, 97%

Chemical Identifiers

Chemical Name or Material 3,5-Dichloropyridine-2-carboxamide
CAS 5468-71-3
Formula Weight 191.01
MDL Number MFCD20529142
Beilstein 136067
Synonym 3,5-Dichloropicolinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6698103

 
alfa aesar™
H6698103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66981MD

 
alfa aesar™
H66981MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.01
Chemical Name or Material 3,5-Dichloropyridine-2-carboxamide
MDL Number MFCD20529142
CAS 5468-71-3
Health Hazard 1 Warning
Physical Form Powder
Synonym 3,5-Dichloropicolinamide
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 136067
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Color Off-white
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C