Learn More
3,5-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1512714
Description
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
3,5-Dichlorophenylhydrazine hydrochloride | |
∼204°C (decomposition) | |
RONG> | |
4208459 | |
Soluble in water. | |
C1=C(C=C(C=C1Cl)Cl)NN.Cl | |
213.486 | |
213.5 |
63352-99-8 | |
C6H7Cl3N2 | |
25 g | |
3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3 | |
BDXZOJVMTJOAPS-UHFFFAOYSA-N | |
(3,5-dichlorophenyl)hydrazine;hydrochloride | |
2723913 | |
97% |
Chemical Identifiers
63352-99-8 | |
213.486 | |
BDXZOJVMTJOAPS-UHFFFAOYSA-N | |
2723913 | |
C1=C(C=C(C=C1Cl)Cl)NN.Cl |
C6H7Cl3N2 | |
RONG> | |
3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3 | |
(3,5-dichlorophenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 264-109-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only