Please login to your online account to display your discounted pricing

3,5-Dichlorophenylhydrazine hydrochloride, 93%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dichlorophenylhydrazine hydrochloride
Name Note 93%
CAS 63352-99-8
CAS Min % 91.0
CAS Max % 100.0
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C6 H6 Cl2 N2 . H Cl
Linear Formula Cl2C6H3NHNH2·HCl
Formula Weight 213.49
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167180050

  SDS
thermo fisher scientific
167180050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥91%
Formula Weight 213.49
Physical Form Powder
Quantity 5g
Color White to light beige
Beilstein 15, I, 116
TSCA TSCA
EINECS Number 264-109-7
CAS Max % 100.0
CAS Min % 91.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
MDL Number MFCD00012938
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Linear Formula Cl2C6H3NHNH2·HCl
Assay Percent Range 93%
CAS 63352-99-8
Name Note 93%
Chemical Name or Material 3, 5-Dichlorophenylhydrazine hydrochloride
Molecular Formula C6 H6 Cl2 N2 . H Cl