Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorophenylhydrazine, 98%

$28.38 - $131.14

Chemical Identifiers

Chemical Name or Material 3,5-Dichlorophenylhydrazine
CAS 39943-56-1
Molecular Formula C6H6Cl2N2
Formula Weight 177.03g/mol
MDL Number MFCD00051527
Beilstein 3126965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1120703

 
alfa aesar™
A11207-03
1g Each for $28.38

AAA1120706

 
alfa aesar™
A11207-06
5g Each for $131.14
Description & Specifications

Specifications

Quantity 1g
Melting Point 112° to 117°C
Quantity 1g
Chemical Name or Material 3,5-Dichlorophenylhydrazine
CAS 39943-56-1
Assay 98%
Molecular Formula C6H6Cl2N2
Formula Weight 177.03g/mol
MDL Number MFCD00051527
Beilstein 3126965
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.