missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorophenylacetic acid, 95%

Manufacturer:  Alfa Aesar™ H63293MD

 View more versions of this product

Catalog No. AAH63293MD



Specifications

Specifications

3,5-Dichlorophenylacetic acid
51719-65-4
MFCD01764671
RERINLRFXYGZEE-UHFFFAOYSA-N
2-(3,5-dichlorophenyl)acetic acid
6452487
Solid
250mg
C8H6Cl2O2
2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid
C1=C(C=C(C=C1Cl)Cl)CC(=O)O
205.034
205.03
95%

Chemical Identifiers

51719-65-4
205.034
RERINLRFXYGZEE-UHFFFAOYSA-N
6452487
C1=C(C=C(C=C1Cl)Cl)CC(=O)O
C8H6Cl2O2
MFCD01764671
2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid
2-(3,5-dichlorophenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents