Please login to your online account to display your discounted pricing

(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (3, 5-Dichlorophenyl)methanesulfonyl chloride
CAS 163295-70-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% as diethylsulfonamide derivative (HPLC)
Assay Percent Range 97%
Molecular Formula C7H5Cl3O2S
Formula Weight 259.54
MDL Number MFCD01631928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC433230010

  SDS
thermo fisher scientific
AC433230010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC433230050

  SDS
thermo fisher scientific
AC433230050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Solid
Formula Weight 259.54
Assay ≥96.0% as diethylsulfonamide derivative (HPLC)
Quantity 1g
Color White to pale yellow
Melting Point 107° to 112°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
MDL Number MFCD01631928
Chemical Name or Material (3, 5-Dichlorophenyl)methanesulfonyl chloride
Molecular Formula C7H5Cl3O2S
CAS 163295-70-3
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.