missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC433230050

 View more versions of this product

Catalog No. AC433230050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(3, 5-Dichlorophenyl)methanesulfonyl chloride
107° to 112°C
163295-70-3
96.0
Powder or Solid
97%
259.54
White to pale yellow
5g
Authentic
100.0
≥96.0% as diethylsulfonamide derivative (HPLC)
C7H5Cl3O2S
MFCD01631928

Chemical Identifiers

163295-70-3
100.0
C7H5Cl3O2S
MFCD01631928
≥96.0% as diethylsulfonamide derivative (HPLC)
96.0
97%
259.54
(3, 5-Dichlorophenyl)methanesulfonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger