Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorophenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dichlorophenyl isothiocyanate
CAS 6590-93-8
Molecular Formula C7H3Cl2NS
Formula Weight 204.08
MDL Number MFCD00041072
UN Number UN2923
Beilstein 879282
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068706

  SDS
alfa aesar™
B2068706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.08
Chemical Name or Material 3,5-Dichlorophenyl isothiocyanate
Odor Irritating
Assay 98%
Molecular Formula C7H3Cl2NS
MDL Number MFCD00041072
CAS 6590-93-8
UN Number UN2923
Boiling Point 137° to 138°C (9mmHg)
Beilstein 879282
Melting Point 44° to 48°C
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.