missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dichlorophenol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113610500
Specifications
3, 5-Dichlorophenol | |
Glass bottle | |
Yellow to Brown | |
50g | |
98.5 | |
98.5% min. (GC) | |
Cl2C6H3OH | |
06, 190 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,dmso | |
C1=C(C=C(C=C1Cl)Cl)O | |
163 | |
163 | |
99% |
Authentic | |
233°C | |
64°C to 68°C | |
591-35-5 | |
100.0 | |
C6H4Cl2O | |
MFCD00002259 | |
3,5 dichlorophenol, 3,5-dichloro phenol, 3,5-dichloro-phenol, acmc-209mar, ccris 5905, dsstox_cid_5006, dsstox_rid_77624, phenol, 3,5-dichloro, pubchem3699, unii-fg32l88ko9 | |
VPOMSPZBQMDLTM-UHFFFAOYSA-N | |
3,5-dichlorophenol | |
11571 | |
Crystals, Crystalline Powder or Crystalline Mass |
Chemical Identifiers
591-35-5 | |
163 | |
VPOMSPZBQMDLTM-UHFFFAOYSA-N | |
11571 | |
C1=C(C=C(C=C1Cl)Cl)O |
C6H4Cl2O | |
MFCD00002259 | |
3,5 dichlorophenol, 3,5-dichloro phenol, 3,5-dichloro-phenol, acmc-209mar, ccris 5905, dsstox_cid_5006, dsstox_rid_77624, phenol, 3,5-dichloro, pubchem3699, unii-fg32l88ko9 | |
3,5-dichlorophenol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 209-714-9