missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorophenol, 98%

Manufacturer:  Alfa Aesar™ A1532818

 View more versions of this product

Catalog No. AAA1532818



Description

Description

Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3,5-Dichlorophenol
233°C
50g
C6H4Cl2O
UN2020
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
C1=C(C=C(C=C1Cl)Cl)O
162.997
163
Phenol-like
63°C to 68°C
591-35-5
MFCD00002259
1906458
VPOMSPZBQMDLTM-UHFFFAOYSA-N
3,5-dichlorophenol
11571
98%

Chemical Identifiers

591-35-5
162.997
VPOMSPZBQMDLTM-UHFFFAOYSA-N
11571
C1=C(C=C(C=C1Cl)Cl)O
C6H4Cl2O
MFCD00002259
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
3,5-dichlorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P273-P305+P351+P338
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

RTECSNumber : SK8820000

TSCA : TSCA

Documents