missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1532806

 View more versions of this product

Catalog No. AAA1532806



Description

Description

3,5-Dichlorophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3,5-Dichlorophenol
63°C to 68°C
Phenol-like
MFCD00002259
UN2020
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
VPOMSPZBQMDLTM-UHFFFAOYSA-N
3,5-dichlorophenol
11571
98%
591-35-5
233°C
C6H4Cl2O
5 g
1906458
Slightly soluble in water. Very soluble in ethanol,ethyl ether and petroleum ether.
C1=C(C=C(C=C1Cl)Cl)O
162.997
163

Chemical Identifiers

591-35-5
162.997
VPOMSPZBQMDLTM-UHFFFAOYSA-N
11571
C1=C(C=C(C=C1Cl)Cl)O
C6H4Cl2O
MFCD00002259
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
3,5-dichlorophenol
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

EINECSNumber : 209-714-9

RTECSNumber : SK8820000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only