missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorobenzylamine, 94%

Manufacturer:  Alfa Aesar™ L1205406

 View more versions of this product

Catalog No. AAL1205406



Specifications

Specifications

3,5-Dichlorobenzylamine
5g
C7H7Cl2N
UN2735
3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
C1=C(C=C(C=C1Cl)Cl)CN
176.04
176.05
141°C to 144°C (15mmHg)
39989-43-0
MFCD00052681
3237459
ICIJWOWQUHHETJ-UHFFFAOYSA-N
(3,5-dichlorophenyl)methanamine
457602
97%

Chemical Identifiers

39989-43-0
176.04
ICIJWOWQUHHETJ-UHFFFAOYSA-N
457602
C1=C(C=C(C=C1Cl)Cl)CN
C7H7Cl2N
MFCD00052681
3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
(3,5-dichlorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents