missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorobenzoyl chloride, 97%, ACROS Organics™

$61.98 - $61.98

Chemical Identifiers

CAS 2905-62-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H3Cl3O
Linear Formula ClC6H3COCl
Formula Weight 209.46
MDL Number MFCD00009817
Chemical Name or Material 3, 5-Dichlorobenzoyl chloride
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC367970050 SDS Thermo Fisher Scientific
AC367970050
Glass bottle 5g Each for $61.98
Specifications Description & Specifications

Chemical Identifiers

2905-62-6
100.0
C7H3Cl3O
209.46
3, 5-Dichlorobenzoyl chloride
96.0
97%
ClC6H3COCl
MFCD00009817
≥96.0%

Specifications

Clear colorless to light yellow
2905-62-6
100.0
Glass bottle
Liquid or Low Melting Solid
C7H3Cl3O
209.46
09, 344
≥96.0%
5g
96.0
Authentic
1.5795 to 1.5825 (20°C, 589nm)
97%
ClC6H3COCl
MFCD00009817
3, 5-Dichlorobenzoyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
In contact with water releases flammable gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger