Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorobenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Dichlorobenzonitrile
CAS 6575-00-4
Molecular Formula C7H3Cl2N
Formula Weight 172.01
MDL Number MFCD00001800
UN Number UN3439
Beilstein 2207019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1121306

  SDS
alfa aesar™
A1121306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.01
Molecular Formula C7H3Cl2N
UN Number UN3439
MDL Number MFCD00001800
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 64° to 68°C
CAS 6575-00-4
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Chemical Name or Material 3,5-Dichlorobenzonitrile
Health Hazard 1 Danger
Assay 98%
Quantity 5g
Beilstein 2207019
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.