Please login to your online account to display your discounted pricing

3,5-Dichlorobenzonitrile, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC160790050

 View more versions of this product

Catalog No. AC160790050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 172.01
Color White to beige-brownish
Quantity 5g
Melting Point 66° to 71°C
Beilstein 09, 345
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3, 5-Dichlorobenzonitrile
CAS 6575-00-4
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
MDL Number MFCD00001800
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C7H3Cl2N
Linear Formula Cl2C6H3CN

Specifications

Chemical Name or Material 3, 5-Dichlorobenzonitrile
CAS 6575-00-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H3Cl2N
Linear Formula Cl2C6H3CN
Formula Weight 172.01
MDL Number MFCD00001800
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents