missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorobenzonitrile, 97%, ACROS Organics™

$46.29 - $46.29

Chemical Identifiers

CAS 6575-00-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H3Cl2N
Linear Formula Cl2C6H3CN
Formula Weight 172.01
MDL Number MFCD00001800
Chemical Name or Material 3, 5-Dichlorobenzonitrile
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC160790050 SDS Thermo Fisher Scientific
AC160790050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC160790010 SDS Thermo Fisher Scientific
160790010
Glass bottle 1g Each for $46.29
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

6575-00-4
100.0
C7H3Cl2N
172.01
3, 5-Dichlorobenzonitrile
96.0
97%
Cl2C6H3CN
MFCD00001800
≥96.0%

Specifications

White to beige-brownish
172.01
66° to 71°C
100.0
6575-00-4
MFCD00001800
Cl2C6H3CN
97%
≥96.0%
Crystalline Powder
5g
09, 345
96.0
Authentic
Glass bottle
C7H3Cl2N
3, 5-Dichlorobenzonitrile

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning