Please login to your online account to display your discounted pricing

3,5-Dichlorobenzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3, 5-Dichlorobenzoic acid
CAS 51-36-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4Cl2O2
Linear Formula Cl2C6H3CO2H
Formula Weight 191.01
MDL Number MFCD00002494
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB03142EA

  SDS
fisher scientific company
BTB03142EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.01
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 3, 5-Dichlorobenzoic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C7H4Cl2O2
Linear Formula Cl2C6H3CO2H
MDL Number MFCD00002494
CAS 51-36-5
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.