missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorobenzhydrazide, 97%

$370.97 - $370.97

Chemical Identifiers

CAS 62899-78-9
Molecular Formula C7H6Cl2N2O
Formula Weight 205.04
MDL Number MFCD00833411
Chemical Name or Material 3,5-Dichlorobenzhydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2238306 SDS Alfa Aesar™
B2238306
5g Each for $370.97
AAB2238303 SDS Alfa Aesar™
B2238303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

62899-78-9
205.04
3,5-Dichlorobenzhydrazide
C7H6Cl2N2O
MFCD00833411

Specifications

5g
C7H6Cl2N2O
MFCD00833411
3,5-Dichlorobenzhydrazide
62899-78-9
205.04
205° to 207°C
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning