missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 705-21-5
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.51
MDL Number MFCD00051698
InChI Key RJSQINMKOSOUGT-UHFFFAOYSA-N
Synonym 3,5-dichloro-benzenesulfonyl chloride
PubChem CID 522388
IUPAC Name 3,5-dichlorobenzenesulfonyl chloride
SMILES C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356530010 SDS View Documents Thermo Fisher Scientific
AC356530010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

705-21-5
245.51
RJSQINMKOSOUGT-UHFFFAOYSA-N
522388
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)Cl
C6H3Cl3O2S
MFCD00051698
3,5-dichloro-benzenesulfonyl chloride
3,5-dichlorobenzenesulfonyl chloride

Specifications

Authentic
Beige
1g
97.5
97.5% min. (GC)
Cl(Cl)C6H3SO2Cl
3,5-dichloro-benzenesulfonyl chloride
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)Cl
245.51
245.51
98%
Glass bottle
53°C to 57°C
705-21-5
100.0
C6H3Cl3O2S
MFCD00051698
RJSQINMKOSOUGT-UHFFFAOYSA-N
3,5-dichlorobenzenesulfonyl chloride
522388
Solid
3, 5-Dichlorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger