Please login to your online account to display your discounted pricing

3,5-Dichlorobenzenesulfonyl chloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC356530010

 View more versions of this product

Catalog No. AC356530010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Solid
Formula Weight 245.51
Quantity 1g
Color Beige
Melting Point 30° to 34°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00051698
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Chemical Name or Material 3, 5-Dichlorobenzenesulfonyl chloride
Assay Percent Range 98%
Molecular Formula C6H3Cl3O2S
Linear Formula Cl(Cl)C6H3SO2Cl
CAS 705-21-5
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dichlorobenzenesulfonyl chloride
CAS 705-21-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3Cl3O2S
Linear Formula Cl(Cl)C6H3SO2Cl
Formula Weight 245.51
MDL Number MFCD00051698
Accessories

GHS Signal Word: Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents