missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorobenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H55923MD

 View more versions of this product

Catalog No. AAH55923MD



Specifications

Specifications

3,5-Dichlorobenzenesulfonamide
250mg
C6H5Cl2NO2S
3,5-bis chloranyl benzenesulfonamide
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)N
226.071
226.08
161°C to 165°C
19797-32-1
MFCD00117161
AHNOVNYOUPQVRX-UHFFFAOYSA-N
3,5-dichlorobenzenesulfonamide
705607
97%

Chemical Identifiers

19797-32-1
226.071
AHNOVNYOUPQVRX-UHFFFAOYSA-N
705607
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)N
C6H5Cl2NO2S
MFCD00117161
3,5-bis chloranyl benzenesulfonamide
3,5-dichlorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

RTECSNumber : DB1670000

TSCA : No

Recommended Storage : Ambient temperatures

Documents