Please login to your online account to display your discounted pricing

3,5-Dichlorobenzene-1-carboximidamide hydrochloride, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 3, 5-Dichlorobenzene-1-carboximidamide hydrochloride
Assay Tech.
Molecular Formula C7H6Cl2N2·ClH
Linear Formula Cl2C6H3C(=NH)NH2·HCl
Formula Weight 225.51
MDL Number MFCD00173785
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB00407DA

  SDS
fisher scientific company
SPB00407DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB00407EA

  SDS
fisher scientific company
SPB00407EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.51
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
Packaging Amber Glass Bottle
Chemical Name or Material 3, 5-Dichlorobenzene-1-carboximidamide hydrochloride
Molecular Formula C7H6Cl2N2·ClH
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00173785
Linear Formula Cl2C6H3C(=NH)NH2·HCl
CAS 22978-61-6
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.