Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichlorobenzamide, 97%

$60.66 - $575.69

Chemical Identifiers

Chemical Name or Material 3,5-Dichlorobenzamide
CAS 5980-23-4
Molecular Formula C7H5Cl2NO
Formula Weight 190.03
MDL Number MFCD00057644
Beilstein 2249768
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2195906

 
alfa aesar™
B2195906
5g Each for $60.66

AAB2195922

 
alfa aesar™
B2195922
100g Each for $575.69
Description & Specifications

Specifications

Formula Weight 190.03
Chemical Name or Material 3,5-Dichlorobenzamide
Assay 97%
Molecular Formula C7H5Cl2NO
MDL Number MFCD00057644
CAS 5980-23-4
Beilstein 2249768
Melting Point 159° to 161°C
Quantity 5g