Please login to your online account to display your discounted pricing

3,5-Dichlorobenzaldehyde 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dichlorobenzaldehyde
Name Note 97%
CAS 10203-08-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4Cl2O
Linear Formula Cl2C6H3CHO
Formula Weight 175.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160800010

  SDS
thermo fisher scientific
160800010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fluffy Powder or Crystalline Mass
Assay ≥96.0%
Formula Weight 175.01
Quantity 1g
Color White to slightly yellow
EINECS Number 233-499-0
TSCA TSCA
Melting Point 61° to 66°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 3, 5-Dichlorobenzaldehyde
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H4Cl2O
Linear Formula Cl2C6H3CHO
MDL Number MFCD00003352
CAS 10203-08-4
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.