Please login to your online account to display your discounted pricing

3,5-Dichloroanthranilic Acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dichloroanthranilic acid
Name Note 97%
CAS 2789-92-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C7 H5 Cl2 N O2
Formula Weight 206.03
MDL Number MFCD00017088
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113131000

  SDS
thermo fisher scientific
113131000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96%
Formula Weight 206.03
Quantity 100g
Color Off-white to yellow to dark pink
EINECS Number 220-520-3
TSCA TSCA
RTECS Number CB2800000
Melting Point 227° to 232°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 2-Amino-3, 5-dichlorobenzoic acid
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 5-Dichloroanthranilic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7 H5 Cl2 N O2
MDL Number MFCD00017088
CAS 2789-92-6
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.