missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3,5-Dichloroanisole, 98%
Manufacturer: Alfa Aesar™ A1552109
Specifications
3,5-Dichloroanisole | |
97°C (8mmHg) | |
10g | |
C7H6Cl2O | |
1936395 | |
SSNXYMVLSOMJLU-UHFFFAOYSA-N | |
1,3-dichloro-5-methoxybenzene | |
36588 | |
98% |
106°C (222°F) | |
39°C to 42°C | |
33719-74-3 | |
MFCD00000589 | |
3,5-dichloroanisole, benzene, 3,5-dichloro-1-methoxy, 3,5-dichloranisol, acmc-20anda, pubchem3684, 3,5-dichloro anisole, 3,5-dichloroanisol, 3,5-dichloroanisole, ssnxymvlsomjlu-uhfffaoysa, # | |
COC1=CC(=CC(=C1)Cl)Cl | |
177.024 | |
177.03 |
Chemical Identifiers
33719-74-3 | |
177.024 | |
SSNXYMVLSOMJLU-UHFFFAOYSA-N | |
36588 | |
COC1=CC(=CC(=C1)Cl)Cl |
C7H6Cl2O | |
MFCD00000589 | |
3,5-dichloroanisole, benzene, 3,5-dichloro-1-methoxy, 3,5-dichloranisol, acmc-20anda, pubchem3684, 3,5-dichloro anisole, 3,5-dichloroanisol, 3,5-dichloroanisole, ssnxymvlsomjlu-uhfffaoysa, # | |
1,3-dichloro-5-methoxybenzene |