Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloroanisole, 98%

$44.45 - $185.34

Chemical Identifiers

Chemical Name or Material 3,5-Dichloroanisole
CAS 33719-74-3
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00000589
Beilstein 1936395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1552109

  SDS
alfa aesar™
A1552109
10g Each for $44.45

AAA1552114

  SDS
alfa aesar™
A1552114
25g Each for $101.09

AAA1552118

  SDS
alfa aesar™
A1552118
50g Each for $185.34
Description & Specifications

Specifications

Formula Weight 177.03
Boiling Point 97°C (8mmHg)
MDL Number MFCD00000589
Molecular Formula C7H6Cl2O
Beilstein 1936395
Chemical Name or Material 3,5-Dichloroanisole
Quantity 10g
Assay 98%
CAS 33719-74-3
Flash Point 106°C (222°F)
Melting Point 39° to 42°C