missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dichloro-N-methylbenzylamine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438870010
Specifications
3, 5-Dichloro-N-methylbenzylamine | |
Glass Bottle | |
1g | |
94.0 | |
94% min. (GC) | |
1-3,5-dichlorophenyl-n-methyl-methanamine, 1-3,5-dichlorophenyl-n-methylmethanamine, 3,5-dichloro-n-methyl-benzenemethanamine, 3,5-dichloro-n-methylbenzylamine, 3,5-dichlorophenyl methyl methyl amine, 90389-22-3 hydrochloride, benzenemethanamine, 3,5-dichloro-n-methyl, benzylamine, n-methyl-3,5-dichloro, n-methyl-3,5-dichlorobenzylamine | |
CNCC1=CC(=CC(=C1)Cl)Cl | |
190.07 | |
190.07 | |
95% |
Authentic | |
Colorless to Yellow | |
90390-21-9 | |
100.0 | |
C8H9Cl2N | |
SSTJQZMMDIIKBR-UHFFFAOYSA-N | |
1-(3,5-dichlorophenyl)-N-methylmethanamine | |
146163 | |
Liquid |
Chemical Identifiers
90390-21-9 | |
190.07 | |
1-3,5-dichlorophenyl-n-methyl-methanamine, 1-3,5-dichlorophenyl-n-methylmethanamine, 3,5-dichloro-n-methyl-benzenemethanamine, 3,5-dichloro-n-methylbenzylamine, 3,5-dichlorophenyl methyl methyl amine, 90389-22-3 hydrochloride, benzenemethanamine, 3,5-dichloro-n-methyl, benzylamine, n-methyl-3,5-dichloro, n-methyl-3,5-dichlorobenzylamine | |
1-(3,5-dichlorophenyl)-N-methylmethanamine |
C8H9Cl2N | |
SSTJQZMMDIIKBR-UHFFFAOYSA-N | |
146163 | |
CNCC1=CC(=CC(=C1)Cl)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger