Learn More
3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5070703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,5-Dichloro-alpha-toluenesulfonyl chloride | |
C7H5Cl3O2S | |
1 g | |
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichlorophenyl methyl sulfonyl chloride, acmc-1c80o, 3,5-dichloro-, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichlorophenyl methane-sulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro | |
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1 | |
259.53 | |
259.54 |
163295-70-3 | |
MFCD01631928 | |
UN3096 | |
GZMJRPMBVICCFL-UHFFFAOYSA-N | |
(3,5-dichlorophenyl)methanesulfonyl chloride | |
4993270 | |
96% |
Chemical Identifiers
163295-70-3 | |
259.53 | |
GZMJRPMBVICCFL-UHFFFAOYSA-N | |
4993270 | |
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1 |
C7H5Cl3O2S | |
MFCD01631928 | |
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichlorophenyl methyl sulfonyl chloride, acmc-1c80o, 3,5-dichloro-, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichlorophenyl methane-sulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro | |
(3,5-dichlorophenyl)methanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only