Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-6-hydroxyaniline, ≥98%

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-6-hydroxyaniline
CAS 527-62-8
Formula Weight 178.01
MDL Number MFCD00035766
UN Number UN2811
Beilstein 1102196
Synonym 2-Amino-4,6-dichlorophenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6480106

 
alfa aesar™
H6480106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6480114

 
alfa aesar™
H6480114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6480122

 
alfa aesar™
H6480122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.01
Melting Point 89° to 94°C
Chemical Name or Material 3,5-Dichloro-6-hydroxyaniline
Physical Form Solid
Assay ≥98%
MDL Number MFCD00035766
CAS 527-62-8
UN Number UN2811
Beilstein 1102196
Synonym 2-Amino-4,6-dichlorophenol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number SJ5774000
Color Yellow to brown
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.