Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-pyridone, 98%

$74.89 - $1,127.05

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-4-pyridone
CAS 17228-70-5
Molecular Formula C5H3Cl2NO
Formula Weight 163.99
MDL Number MFCD00051673
Beilstein 1524978
Synonym 3,5-Dichloro-4-hydroxypyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1424709

 
alfa aesar™
A1424709
10g Each for $74.89

AAA1424718

 
alfa aesar™
A1424718
50g Each for $268.20

AAA1424730

 
alfa aesar™
A1424730
250g Each for $1,127.05
Description & Specifications

Specifications

Formula Weight 163.99
Chemical Name or Material 3,5-Dichloro-4-pyridone
Assay 98%
Molecular Formula C5H3Cl2NO
MDL Number MFCD00051673
CAS 17228-70-5
Beilstein 1524978
Synonym 3,5-Dichloro-4-hydroxypyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 320° to 325°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.