Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-pyridone-1-acetic acid, 98%

$93.43 - $367.89

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-4-pyridone-1-acetic acid
CAS 56187-37-2
Molecular Formula C7H5Cl2NO3
Formula Weight 222.03
MDL Number MFCD02180547
Synonym 3,5-Dichloro-4-oxo-1(4H)-pyridineacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2547106

 
alfa aesar™
B2547106
5g Each for $93.43

AAB2547114

 
alfa aesar™
B2547114
25g Each for $367.89
Description & Specifications

Specifications

Formula Weight 222.03
Chemical Name or Material 3,5-Dichloro-4-pyridone-1-acetic acid
Assay 98%
Molecular Formula C7H5Cl2NO3
MDL Number MFCD02180547
CAS 56187-37-2
Melting Point 239° to 241°C
Synonym 3,5-Dichloro-4-oxo-1(4H)-pyridineacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g

Applications
3,5-Dichloro-4-pyridone-1-acetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.