Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-hydroxybenzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-4-hydroxybenzenesulfonyl chloride
CAS 13432-81-0
Molecular Formula C6H3Cl3O3S
Formula Weight 261.51
MDL Number MFCD00171551
UN Number UN3261
Beilstein 2806977
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2017403

  SDS
alfa aesar™
L2017403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.51
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzenesulfonyl chloride
CAS 13432-81-0
MDL Number MFCD00171551
Molecular Formula C6H3Cl3O3S
UN Number UN3261
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Beilstein 2806977
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 118° to 119°C
Quantity 1g
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is employed in the convergent syntheses involving hydroxyarenesulfonyl chlorides.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.