Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-fluorobenzotrifluoride, 98%

$51.08 - $159.72

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-4-fluorobenzotrifluoride
CAS 77227-81-7
Molecular Formula C7H2Cl2F4
Formula Weight 232.99
MDL Number MFCD00068183
Beilstein 6199573
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1849703

  SDS
alfa aesar™
L1849703
1g Each for $51.08

AAL1849706

  SDS
alfa aesar™
L1849706
5g Each for $159.72
Description & Specifications

Specifications

Formula Weight 232.99
Chemical Name or Material 3,5-Dichloro-4-fluorobenzotrifluoride
Assay 98%
MDL Number MFCD00068183
Health Hazard 1 Warning
Beilstein 6199573
Molecular Formula C7H2Cl2F4
CAS 77227-81-7
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 73°C (163°F)
Boiling Point 166° to 168°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.